missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-(2-oxopropyl)-2,4,7-trioxabicyclo[3.3.0]octane 95.0+%, TCI America™

Catalog No. m19785g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19785G Supplier TCI America Supplier No. M19785G

Chemical Identifiers

CAS 1378825-86-5
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
InChI Key RGYHEPWCPQAJSI-UHFFFAOYSA-N
PubChem CID 44629945
IUPAC Name 1-(2-methyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-2-yl)propan-2-one
SMILES CC(=O)CC1(OC2COCC2O1)C

Specifications

Melting Point 56°C
Color White-Yellow
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-3-(2-oxopropyl)-2,4,7-trioxabicyclo[3.3.0]octane