missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. M14701G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14701G Supplier TCI America Supplier No. M14701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55589-47-4
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD02181144
InChI Key JDYVLWWFVYNMTN-UHFFFAOYSA-N
Synonym 3-methylpicolinaldehyde, 3-methyl-2-pyridinecarboxaldehyde, 3-methylpyridine-2-carboxaldehyde, 3-methyl-2-pyridinaldehyde, 2-formyl-3-picoline, 3-methyl-pyridine-2-carbaldehyde, 2-formyl-3-methylpyridine, pyridinecarboxaldehyde, methyl, 2-pyridinecarboxaldehyde, 3-methyl, 3-methyl-2-pyridineformaldehyde
PubChem CID 1501874
IUPAC Name 3-methylpyridine-2-carbaldehyde
SMILES CC1=C(N=CC=C1)C=O

Specifications

Melting Point 8°C
Color Yellow
Boiling Point 84°C
Quantity 1 g
Formula Weight 121.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-pyridinecarboxaldehyde