missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-pentanone 98.0+%, TCI America™
Supplier: TCI America M038825ML
Specifications
| 3-Methyl-2-pentanone | |
| Yellow | |
| C6H12O | |
| 25 mL | |
| 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4 | |
| CCC(C)C(=O)C | |
| 100.161 | |
| 100.16 | |
| Liquid |
| 565-61-7 | |
| 116°C | |
| MFCD00009336 | |
| 1224 | |
| UIHCLUNTQKBZGK-UHFFFAOYSA-N | |
| 3-methylpentan-2-one | |
| 11262 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 565-61-7 | |
| 100.161 | |
| UIHCLUNTQKBZGK-UHFFFAOYSA-N | |
| 11262 | |
| CCC(C)C(=O)C |
| C6H12O | |
| MFCD00009336 | |
| 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4 | |
| 3-methylpentan-2-one |
Safety and Handling
EINECSNumber : (2)-0542
TSCA : No