missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pentanone 98.0+%, TCI America™

Catalog No. M03885ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03885ML 5 mL
M038825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M03885ML Supplier TCI America Supplier No. M03885ML

Chemical Identifiers

CAS 565-61-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00009336
InChI Key UIHCLUNTQKBZGK-UHFFFAOYSA-N
Synonym 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
PubChem CID 11262
IUPAC Name 3-methylpentan-2-one
SMILES CCC(C)C(=O)C

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 mL
UN Number 1224
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-pentanone
EINECSNumber (2)-0542
TSCA No