missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pentanone, 99%

CAS: 565-61-7 | C6H12O | 100.16 g/mol

$132.14 - $132.14

Chemical Identifiers

CAS 565-61-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
InChI Key UIHCLUNTQKBZGK-UHFFFAOYSA-N
Synonym 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
PubChem CID 11262
IUPAC Name 3-methylpentan-2-one
SMILES CCC(C)C(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127380050
SDS View Documents
Thermo Scientific Chemicals
127380050
5 g Glass bottle
Each for $132.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

565-61-7
100.16
3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
3-methylpentan-2-one
C6H12O
UIHCLUNTQKBZGK-UHFFFAOYSA-N
11262
CCC(C)C(=O)C

Specifications

565-61-7
100.0
0.8100g/mL
12°C
98.5% min. (GC)
C6H12O
C2H5CH(CH3)COCH3
01,693
3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
CCC(C)C(=O)C
100.16
100.16
Liquid
98.5
Colorless
118.0°C (758.0 mmHg)
Authentic
Glass bottle
1.4002
5 g
0.81
UIHCLUNTQKBZGK-UHFFFAOYSA-N
3-methylpentan-2-one
11262
99%
3-Methyl-2-pentanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapour.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-282-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only