Learn More
3-Methyl-2-pentanone, 98+%
CAS: 565-61-7 | C6H12O | 100.161 g/mol
$176.53 - $619.62
Chemical Identifiers
| CAS | 565-61-7 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.161 |
| MDL Number | MFCD00009336 |
| InChI Key | UIHCLUNTQKBZGK-UHFFFAOYSA-N |
| Synonym | 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4 |
| PubChem CID | 11262 |
| IUPAC Name | 3-methylpentan-2-one |
| SMILES | CCC(C)C(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1999909
|
Thermo Scientific Chemicals
A1999909 |
10 g |
N/A
|
|
|||||
|
AAA1999914
|
Thermo Scientific Chemicals
A1999914 |
25 g |
N/A
|
|
|||||
|
AAA1999918
|
Thermo Scientific Chemicals
A1999918 |
50 g |
N/A
|
|
|||||
Description
3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.
Solubility
Soluble in alcohol.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Chemical Identifiers
| 565-61-7 | |
| 100.161 | |
| UIHCLUNTQKBZGK-UHFFFAOYSA-N | |
| 11262 | |
| CCC(C)C(=O)C |
| C6H12O | |
| MFCD00009336 | |
| 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4 | |
| 3-methylpentan-2-one |
Specifications
| 565-61-7 | |
| 118°C to 119°C | |
| C6H12O | |
| MFCD00009336 | |
| UN1224 | |
| 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4 | |
| UIHCLUNTQKBZGK-UHFFFAOYSA-N | |
| 3-methylpentan-2-one | |
| 11262 | |
| ≥98% |
| 0.815 | |
| 12°C (53°F) | |
| 1.4 00 | |
| 10 g | |
| 635791 | |
| Soluble in alcohol. | |
| CCC(C)C(=O)C | |
| 100.161 | |
| 100.16 | |
| 3-Methyl-2-pentanone |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 209-282-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only