missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-pentanol (mixture of diastereoisomers) 95.0+%, TCI America™
$190.13 - $559.22
Chemical Identifiers
| CAS | 565-60-6 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.177 |
| MDL Number | MFCD00004528 |
| InChI Key | ZXNBBWHRUSXUFZ-UHFFFAOYSA-N |
| Synonym | 3-methyl-2-pentanol, 3-methyl-4-pentanol, 2-pentanol, 3-methyl, 2-hydroxy-3-methylpentane, threo-3-methylpentan-2-ol, 2-pentanol, 3-methyl-, 2s,3r, 3-methyl-pentan-2-ol, acmc-1aozb, acmc-20m2p7, sec-butyl methylcarbinol |
| PubChem CID | 11261 |
| ChEBI | CHEBI:77520 |
| IUPAC Name | 3-methylpentan-2-ol |
| SMILES | CCC(C)C(C)O |
Chemical Identifiers
| 565-60-6 | |
| 102.177 | |
| ZXNBBWHRUSXUFZ-UHFFFAOYSA-N | |
| 11261 | |
| 3-methylpentan-2-ol |
| C6H14O | |
| MFCD00004528 | |
| 3-methyl-2-pentanol, 3-methyl-4-pentanol, 2-pentanol, 3-methyl, 2-hydroxy-3-methylpentane, threo-3-methylpentan-2-ol, 2-pentanol, 3-methyl-, 2s,3r, 3-methyl-pentan-2-ol, acmc-1aozb, acmc-20m2p7, sec-butyl methylcarbinol | |
| CHEBI:77520 | |
| CCC(C)C(C)O |
Specifications
| 565-60-6 | |
| C6H14O | |
| 5 mL | |
| 3-methyl-2-pentanol, 3-methyl-4-pentanol, 2-pentanol, 3-methyl, 2-hydroxy-3-methylpentane, threo-3-methylpentan-2-ol, 2-pentanol, 3-methyl-, 2s,3r, 3-methyl-pentan-2-ol, acmc-1aozb, acmc-20m2p7, sec-butyl methylcarbinol | |
| CCC(C)C(C)O | |
| 102.177 | |
| CHEBI:77520 | |
| ≥95.0% (GC) | |
| 3-Methyl-2-pentanol (mixture of diastereoisomers) |
| Colorless | |
| MFCD00004528 | |
| 1987 | |
| ZXNBBWHRUSXUFZ-UHFFFAOYSA-N | |
| 3-methylpentan-2-ol | |
| 11261 | |
| 102.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No