missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pentanol (mixture of diastereoisomers) 95.0+%, TCI America™

Catalog No. M08505ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08505ML 5 mL
M085025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08505ML Supplier TCI America Supplier No. M08505ML
Only null left

Chemical Identifiers

CAS 565-60-6
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004528
InChI Key ZXNBBWHRUSXUFZ-UHFFFAOYSA-N
Synonym 3-methyl-2-pentanol, 3-methyl-4-pentanol, 2-pentanol, 3-methyl, 2-hydroxy-3-methylpentane, threo-3-methylpentan-2-ol, 2-pentanol, 3-methyl-, 2s,3r, 3-methyl-pentan-2-ol, acmc-1aozb, acmc-20m2p7, sec-butyl methylcarbinol
PubChem CID 11261
ChEBI CHEBI:77520
IUPAC Name 3-methylpentan-2-ol
SMILES CCC(C)C(C)O

Specifications

Color Colorless
Quantity 5 mL
UN Number 1987
Formula Weight 102.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-pentanol (mixture of diastereoisomers)
EINECSNumber (2)-0217
TSCA No