missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-oxovaleric Acid 95.0+%, TCI America™

Catalog No. K00201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00201G 1 g
K00205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00201G Supplier TCI America Supplier No. K00201G

Chemical Identifiers

CAS 1460-34-0
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00002582
InChI Key JVQYSWDUAOAHFM-UHFFFAOYSA-N
Synonym 3-Methyl-2-oxopentanoic Acid
PubChem CID 47
ChEBI CHEBI:35932
IUPAC Name 3-methyl-2-oxopentanoic acid
SMILES CCC(C)C(=O)C(=O)O

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 84°C
Quantity 1 g
Formula Weight 130.14
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-2-oxovaleric Acid
EINECSNumber (2)-1503
TSCA No
Recommended Storage Refrigerator