missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 247.66 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Methyl-2-oxo-2, 3-dihydro-1, 3-benzoxazole-6-sulfonyl chloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes serious eye damage. Harmful if swallowed. Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |