missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-oxazolidone 99.0+%, TCI America™

Catalog No. M078125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M078125G 25 g
M0781100G 100 g
M0781500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M078125G Supplier TCI America Supplier No. M078125G
Only null left

Chemical Identifiers

CAS 19836-78-3
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD00009759
InChI Key VWIIJDNADIEEDB-UHFFFAOYSA-N
Synonym 2-Oxo-3-methyloxazolidine
PubChem CID 29831
ChEBI CHEBI:39997
IUPAC Name 3-methyl-1,3-oxazolidin-2-one
SMILES CN1CCOC1=O

Specifications

Boiling Point 88°C
Color Yellow
Quantity 25 g
Formula Weight 101.11
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-oxazolidone
EINECSNumber (5)-5494
RTECSNumber RQ3550000
TSCA Yes