missing translation for 'onlineSavingsMsg'
Learn More

(3-Methyl-2-furyl)methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC55513DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC55513DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

388072-09-1
111.144
CTGIVQJZUKJDNH-UHFFFAOYSA-N
18187367
CC1=C(OC=C1)CN
C6H9NO
MFCD09811371
3-methylfuran-2-yl methanamine, 3-methyl-2-furyl methylamine, 2-aminomethyl-3-methylfuran, 2-furanmethanamine,3-methyl, 2-furanmethanamine, 3-methyl, 1-3-methylfuran-2-yl methanamine, 3-methylfurfurylamine, 3-methylfur-2-yl methylamine
(3-methylfuran-2-yl)methanamine

Specifications

(3-Methyl-2-furyl)methylamine
96.5
97%
C6H9NO
MFCD09811371
CTGIVQJZUKJDNH-UHFFFAOYSA-N
(3-methylfuran-2-yl)methanamine
18187367
97%
388072-09-1
100.0
Amber Glass Bottle
1g
3-methylfuran-2-yl methanamine, 3-methyl-2-furyl methylamine, 2-aminomethyl-3-methylfuran, 2-furanmethanamine,3-methyl, 2-furanmethanamine, 3-methyl, 1-3-methylfuran-2-yl methanamine, 3-methylfurfurylamine, 3-methylfur-2-yl methylamine
CC1=C(OC=C1)CN
111.144
111.14
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

Promotions

Promotions