missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclopenten-1-one, 98%, stabilized

Catalog No. AC163590100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163590100 10 g Glass bottle
AC163590500 50 g Glass bottle
AC163592500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163590100 Supplier Thermo Scientific Chemicals Supplier No. 163590100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2758-18-1 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2758-18-1
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
InChI Key CHCCBPDEADMNCI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one
PubChem CID 17691
IUPAC Name 3-methylcyclopent-2-en-1-one
SMILES CC1=CC(=O)CC1

Specifications

Melting Point 3.0°C to 5.0°C
Color Brown-Yellow to Yellow
Density 0.9800g/mL
Boiling Point 74.0°C (15.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C5H5(=O)
Refractive Index 1.4870 to 1.4900
Quantity 10 g
Beilstein 07,I,46
Specific Gravity 0.98
Solubility Information Solubility in water: miscible
Stabilizer 0.1% hydroquinone
Formula Weight 96.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-cyclopenten-1-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-421-5

RUO – Research Use Only