missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclopenten-1-one, 98%, stab. with 0.1% hydroquinone

Catalog No. AAA1399309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1399309 10 g
AAA1399318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1399309 Supplier Thermo Scientific Chemicals Supplier No. A1399309

CAS: 2758-18-1 | C6H8O | 96.129 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, heat. Store in refrigerator.

Chemical Identifiers

CAS 2758-18-1
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00001403
InChI Key CHCCBPDEADMNCI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one
PubChem CID 17691
IUPAC Name 3-methylcyclopent-2-en-1-one
SMILES CC1=CC(=O)CC1

Specifications

Melting Point 2°C
Density 0.98
pH 4.9
Boiling Point 72°C to 74°C (15 mmHg)
Flash Point 65°C (149°F)
Odor Pleasant
Refractive Index 1.489
Quantity 10 g
Beilstein 1280476
Solubility Information Fully miscible in water.
Formula Weight 96.13
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-cyclopenten-1-one, stabilized with 0.1% hydroquinone
Show More Show Less
Hazard Category H227-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
EINECSNumber 220-421-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only