missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclohexen-1-one, 98%

CAS: 1193-18-6 | C7H10O | 110.16 g/mol

Supplier:  Thermo Scientific Chemicals A1570418

 View more versions of this product

Catalog No. AAA1570418


Only null left
Explore available promotions

Description

Description

3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
Specifications

Specifications

3-Methyl-2-cyclohexen-1-one
0.971
68°C (154°F)
C7H10O
MFCD00001581
1560601
Soluble in alcohol. Insoluble in water.
CC1=CC(=O)CCC1
110.16
110.15
1193-18-6
199°C to 200°C
Pleasant
1.494
50 g
3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
IITQJMYAYSNIMI-UHFFFAOYSA-N
3-methylcyclohex-2-en-1-one
14511
98%

Chemical Identifiers

1193-18-6
110.16
IITQJMYAYSNIMI-UHFFFAOYSA-N
14511
CC1=CC(=O)CCC1
C7H10O
MFCD00001581
3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
3-methylcyclohex-2-en-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c

H227-H302

EINECSNumber : 214-769-7

RTECSNumber : GW7340000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only