missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-butenyl Acetate 98.0+%, TCI America™
$42.87 - $42.87
Chemical Identifiers
| CAS | 1191-16-8 |
|---|---|
| Molecular Formula | C7H12O2 |
| Molecular Weight (g/mol) | 128.17 |
| MDL Number | MFCD00036569 |
| InChI Key | XXIKYCPRDXIMQM-UHFFFAOYSA-N |
| Synonym | prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate |
| PubChem CID | 14489 |
| IUPAC Name | 3-methylbut-2-en-1-yl acetate |
| SMILES | CC(C)=CCOC(C)=O |
Chemical Identifiers
| 1191-16-8 | |
| 128.17 | |
| XXIKYCPRDXIMQM-UHFFFAOYSA-N | |
| 14489 | |
| CC(C)=CCOC(C)=O |
| C7H12O2 | |
| MFCD00036569 | |
| prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate | |
| 3-methylbut-2-en-1-yl acetate |
Specifications
| 1191-16-8 | |
| 128°C | |
| MFCD00036569 | |
| 3272 | |
| XXIKYCPRDXIMQM-UHFFFAOYSA-N | |
| 3-methylbut-2-en-1-yl acetate | |
| 14489 | |
| ≥98.0% (GC) | |
| 3-Methyl-2-butenyl Acetate |
| Colorless | |
| C7H12O2 | |
| 25 mL | |
| prenyl acetate, 3,3-dimethylallyl acetate, 3-methyl-2-butenyl acetate, isopent-2-enyl acetate, 3-methylbut-2-en-1-yl acetate, dimethylallyl acetate, 2-buten-1-ol, 3-methyl-, acetate, 3-methyl-2-buten-1-ol, acetate, unii-i7kov03hgs, isopentenyl acetate | |
| CC(C)=CCOC(C)=O | |
| 128.17 | |
| 128.17 | |
| Liquid |
Safety and Handling
RTECSNumber : EM9473700
TSCA : Yes