missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-buten-1-ol 98.0+%, TCI America™

Catalog No. p-7132316
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
M071425ML 25 mL
M0714100ML 100 mL
M0714500ML 500 mL
3 options

Catalog No. M071425ML

Supplier: TCI America M071425ML

Only null left

Chemical Identifiers

CAS 556-82-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002916
InChI Key ASUAYTHWZCLXAN-UHFFFAOYSA-N
Synonym 3-methyl-2-buten-1-ol, prenol, 3,3-dimethylallyl alcohol, 2-buten-1-ol, 3-methyl, prenyl alcohol, 3-methyl-2-butenol, dimethylallyl alcohol, 3-methyl-2-butenyl alcohol, 3-methylcrotyl alcohol, 3-methyl-2-butene-1-ol
PubChem CID 11173
ChEBI CHEBI:16019
IUPAC Name 3-methylbut-2-en-1-ol
SMILES CC(C)=CCO

Specifications

Color Colorless
Boiling Point 140°C
Quantity 25 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-buten-1-ol
EINECSNumber (2)-2393
RTECSNumber EM9472500
TSCA Yes