missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-buten-1-ol, 98+%

Catalog No. aaa1608914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1608914 25 g
AAA1608922 100 g
AAA1608936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1608914 Supplier Thermo Scientific Chemicals Supplier No. A1608914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 556-82-1 | C5H10O | 86.13 g/mol

3-Methyl-2-buten-1-ol is used as a reagent for protection of carboxylic acids as their 3-methyl-2-buten-1-yl (Prenyl) esters. It is also used in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-buten-1-ol is used as a reagent for protection of carboxylic acids as their 3-methyl-2-buten-1-yl (Prenyl) esters. It is also used in perfumery.

Solubility
Soluble in water (170 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 556-82-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002916
InChI Key ASUAYTHWZCLXAN-UHFFFAOYSA-N
Synonym 3-methyl-2-buten-1-ol, prenol, 3,3-dimethylallyl alcohol, 2-buten-1-ol, 3-methyl, prenyl alcohol, 3-methyl-2-butenol, dimethylallyl alcohol, 3-methyl-2-butenyl alcohol, 3-methylcrotyl alcohol, 3-methyl-2-butene-1-ol
PubChem CID 11173
ChEBI CHEBI:16019
IUPAC Name 3-methylbut-2-en-1-ol
SMILES CC(C)=CCO

Specifications

Density 0.86
Boiling Point 139°C to 140°C
Flash Point 43°C (109°F)
Refractive Index 1.444
Quantity 25 g
UN Number UN1987
Beilstein 1633479
Solubility Information Soluble in water (170g/L at 20°C).
Formula Weight 86.13
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-buten-1-ol
Show More Show Less
Hazard Category H226-H302-H314-H317
Hazard Statement GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 209-141-4
RTECSNumber EM9472500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only