missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone 99.0+%, TCI America™

Catalog No. p-7132035
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M017325ML 25 mL
M0173500ML 500 mL
2 options

Catalog No. M017325ML

Supplier: TCI America M017325ML

Only null left

Chemical Identifiers

CAS 563-80-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00008919
InChI Key SYBYTAAJFKOIEJ-UHFFFAOYSA-N
Synonym 3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
PubChem CID 11251
IUPAC Name 3-methylbutan-2-one
SMILES CC(C)C(C)=O

Specifications

Melting Point -92°C
Color Colorless
Boiling Point 93°C
Quantity 25 mL
UN Number 2397
Formula Weight 86.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butanone
EINECSNumber (2)-0542
RTECSNumber EL9100000
TSCA Yes