missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 218770051

Catalog No. AC218770051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Methyl-2-butanone
97.5
0.8000g/mL
6°C
97.5% min. (GC)
C5H10O
(CH3)2CHCOCH3
5L
0.8
3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
SYBYTAAJFKOIEJ-UHFFFAOYSA-N
3-methylbutan-2-one
11251
98%
563-80-4
100.0
94.0°C to 95.0°C
Authentic
Plastic drum
1.3870 to 1.3890
MFCD00008919
01, 682
15, 6162
Solubility in water: 6g/L (20 c). Other solubilities: miscible with most common organic solvents
CC(C)C(C)=O
86.13
86.13
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-264-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only