missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone, 98%

Catalog No. p-7027628
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB24527AK 250 mL
AAB24527AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB24527AK Supplier Thermo Scientific Chemicals Supplier No. B24527AK
Only null left

CAS: 563-80-4 | C5H10O | 86.13 g/mol

3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-butanone is used for synthesis of heterocyclic compounds and important intermediate of medicine. It is used as an industrial solvent.

Solubility
Soluble in water (8.21 mg/ml at 20°C), ether, alcohol, acetone, and ethanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 563-80-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00008919
InChI Key SYBYTAAJFKOIEJ-UHFFFAOYSA-N
Synonym 3-methyl-2-butanone, methyl isopropyl ketone, isopropyl methyl ketone, 2-acetylpropane, 2-butanone, 3-methyl, methyl butanone-2, mipk, 3-methylbutanone, ketone, isopropyl methyl, 2-methyl-3-butanone
PubChem CID 11251
IUPAC Name 3-methylbutan-2-one
SMILES CC(C)C(C)=O

Specifications

Melting Point -92°C
Density 0.805
Boiling Point 94°C to 95°C
Flash Point −1°C (30°F)
Odor Fruit-like
Refractive Index 1.388
Quantity 250 mL
UN Number UN2397
Beilstein 1071238
Merck Index 14,6090
Solubility Information Soluble in water (8.21mg/ml at 20°C),ether,alcohol,acetone,and ethanol.
Formula Weight 86.13
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-butanone
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 3-METHYLBUTAN-2-ONE
EINECSNumber 209-264-3
RTECSNumber EL9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only