missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinonehydrazone Hydrochloride 98.0+%, TCI America™

Catalog No. p-7132025
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M01605G 5 g
M016025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M01605G Supplier TCI America Supplier No. M01605G
Only null left

Chemical Identifiers

CAS 4338-98-1
Molecular Formula C8H10ClN3S
Molecular Weight (g/mol) 215.699
MDL Number MFCD00005816
InChI Key OEZPVSPULCMUQB-DQMXGCRQSA-N
PubChem CID 9575839
IUPAC Name (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride
SMILES CN1C2=CC=CC=C2SC1=NN.Cl

Specifications

Quantity 5 g
UN Number 2811
Synonym MBTH Hydrochloride, Sawicki′Cs Reagent
Formula Weight 215.70
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-2-benzothiazolinonehydrazone Hydrochloride
RTECSNumber DL7160200
TSCA Yes