Learn More
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
CAS: 149022-15-1 | C8H10ClN3S | 215.70 g/mol
Supplier: Thermo Scientific Chemicals A1631714
Description
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.
Solubility
Soluble in water, dimethyl sulfoxide and methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate | |
| ∼275°C (decomposition) | |
| MFCD00149370 | |
| UN2811 | |
| unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride | |
| OEZPVSPULCMUQB-UHFFFAOYSA-N | |
| (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride | |
| 9575839 | |
| 97% |
| 149022-15-1 | |
| C8H10ClN3S | |
| 25 g | |
| 3571041 | |
| Soluble in water,dimethyl sulfoxide and methanol. | |
| Cl.CN1C(SC2=CC=CC=C12)=NN | |
| 215.70 | |
| 215.71 (Anhydrous) |
Chemical Identifiers
| 149022-15-1 | |
| 215.70 | |
| OEZPVSPULCMUQB-UHFFFAOYSA-N | |
| 9575839 | |
| Cl.CN1C(SC2=CC=CC=C12)=NN |
| C8H10ClN3S | |
| MFCD00149370 | |
| unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride | |
| (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 238-428-7
RTECSNumber : DL7160200
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only