missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2,4-pentanedione 96.0+%, TCI America™

Catalog No. p-7132553
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11325ML 5 mL
M113225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11325ML Supplier TCI America Supplier No. M11325ML
Only null left

Chemical Identifiers

CAS 815-57-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00008762
InChI Key GSOHKPVFCOWKPU-UHFFFAOYSA-N
Synonym 3-methyl-2,4-pentanedione, methylacetylacetone, 1,1-diacetylethane, 2,4-pentanedione, 3-methyl, methyl acetylacetone, 3-acetylbutan-2-one, 3-methyl-acetylacetone, acmc-209plp, 3-methyl-2,4-pentandione, 3-methylpentan-2,4-dione
PubChem CID 69949
IUPAC Name 3-methylpentane-2,4-dione
SMILES CC(C(=O)C)C(=O)C

Specifications

Color Yellow
Boiling Point 172°C
Quantity 5 mL
UN Number 1224
Formula Weight 114.14
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2,4-pentanedione
TSCA No
Recommended Storage Refrigerator