missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1H-pyrazole, 97%

CAS: 1453-58-3 | C4H6N2 | 82.11 g/mol

Supplier:  Thermo Scientific Chemicals L0161814

 View more versions of this product

Catalog No. AAL0161814

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methyl-1H-pyrazole
1.02
93°C (199°F)
1.496
25 g
3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u
CC1=CC=NN1
82.11
82.11
1453-58-3
203°C to 205°C
C4H6N2
MFCD00005240,MFCD08685900
1454
XKVUYEYANWFIJX-UHFFFAOYSA-N
5-methyl-1H-pyrazole
15073
97%

Chemical Identifiers

1453-58-3
82.11
XKVUYEYANWFIJX-UHFFFAOYSA-N
5-methyl-1H-pyrazole
C4H6N2
MFCD00005240,MFCD08685900
15073
CC1=CC=NN1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H335

EINECSNumber : 215-925-7

RTECSNumber : UQ7340000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only