missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1H-pyrazole, 97%

Catalog No. AAL0161806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0161806 5 g
AAL0161814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0161806 Supplier Thermo Scientific Chemicals Supplier No. L0161806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1453-58-3 | C4H6N2 | 82.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1453-58-3
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005240,MFCD08685900
InChI Key XKVUYEYANWFIJX-UHFFFAOYSA-N
PubChem CID 15073
IUPAC Name 5-methyl-1H-pyrazole
SMILES CC1=CC=NN1

Specifications

Density 1.02
Boiling Point 203°C to 205°C
Flash Point 93°C (199°F)
Refractive Index 1.496
Quantity 5 g
Beilstein 1454
Synonym 3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u
Formula Weight 82.11
Percent Purity 97%
Chemical Name or Material 3-Methyl-1H-pyrazole
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 215-925-7
RTECSNumber UQ7340000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only