missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1H-pyrazole-5-carboxylic acid, 97%

Catalog No. p-7040196
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5989903 1 g
AAH5989906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5989903 Supplier Thermo Scientific Chemicals Supplier No. H5989903

CAS: 402-61-9 | C5H6N2O2 | 126.115 g/mol

3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 402-61-9
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00462235
InChI Key WSMQKESQZFQMFW-UHFFFAOYSA-N
Synonym 3-methyl-1h-pyrazole-5-carboxylic acid, 5-methylpyrazole-3-carboxylic acid, 5-methyl-2h-pyrazole-3-carboxylic acid, 3-methylpyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-methyl, 5-methylpyrazole-3-carboxylicacid, 3-methyl-1h-pyrazole-5-carboxylicacid, pyrazole-3-carboxylic acid, 5-methyl, pubchem7842, 5-methyl-2h-pyrazole-3-carboxylicacid
PubChem CID 9822
ChEBI CHEBI:74739
IUPAC Name 5-methyl-1H-pyrazole-3-carboxylic acid
SMILES CC1=CC(=NN1)C(=O)O

Specifications

Melting Point 236°C to 240°C
Quantity 1 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 3-Methyl-1H-pyrazole-5-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UQ6407500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only