Learn More
3-Methyl-1H-pyrazole-5-carboxylic acid, 97%
CAS: 402-61-9 | C5H6N2O2 | 126.115 g/mol
Supplier: Thermo Scientific Chemicals H5989906
Description
3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
| 3-Methyl-1H-pyrazole-5-carboxylic acid | |
| 236°C to 240°C | |
| MFCD00462235 | |
| 3-methyl-1h-pyrazole-5-carboxylic acid, 5-methylpyrazole-3-carboxylic acid, 5-methyl-2h-pyrazole-3-carboxylic acid, 3-methylpyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-methyl, 5-methylpyrazole-3-carboxylicacid, 3-methyl-1h-pyrazole-5-carboxylicacid, pyrazole-3-carboxylic acid, 5-methyl, pubchem7842, 5-methyl-2h-pyrazole-3-carboxylicacid | |
| WSMQKESQZFQMFW-UHFFFAOYSA-N | |
| 5-methyl-1H-pyrazole-3-carboxylic acid | |
| 9822 | |
| 126.11 |
| 402-61-9 | |
| C5H6N2O2 | |
| 5 g | |
| Soluble in dimethyl sulfoxide. | |
| CC1=CC(=NN1)C(=O)O | |
| 126.115 | |
| CHEBI:74739 | |
| 97% |
Chemical Identifiers
| 402-61-9 | |
| 126.115 | |
| WSMQKESQZFQMFW-UHFFFAOYSA-N | |
| 9822 | |
| 5-methyl-1H-pyrazole-3-carboxylic acid |
| C5H6N2O2 | |
| MFCD00462235 | |
| 3-methyl-1h-pyrazole-5-carboxylic acid, 5-methylpyrazole-3-carboxylic acid, 5-methyl-2h-pyrazole-3-carboxylic acid, 3-methylpyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-methyl, 5-methylpyrazole-3-carboxylicacid, 3-methyl-1h-pyrazole-5-carboxylicacid, pyrazole-3-carboxylic acid, 5-methyl, pubchem7842, 5-methyl-2h-pyrazole-3-carboxylicacid | |
| CHEBI:74739 | |
| CC1=CC(=NN1)C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : UQ6407500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only