missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenylindolin-2-one, 97%

CAS: 23210-22-2 | C15H13NO | 223.275 g/mol

$188.89 - $778.49

Chemical Identifiers

CAS 23210-22-2
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD00798609
InChI Key DJKASMOURATDIE-UHFFFAOYSA-N
Synonym 3-methyl-1-phenylindolin-2-one, 3-methyl-1-phenylindoline-2-one, 3-methyl-1-phenyl-2,3-dihydro-1h-indol-2-one, acmc-1ciq4, 1-phenyl-3-methylindoline-2-one, 3-methyl-1-phenyl-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one,1,3-dihydro-3-methyl-1-phenyl
PubChem CID 4990835
IUPAC Name 3-methyl-1-phenyl-3H-indol-2-one
SMILES CC1C2=CC=CC=C2N(C1=O)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4380803
View Documents
Thermo Scientific Chemicals
04380803
1 g
Each for $188.89
Only null left
 
AA4380806
View Documents
Thermo Scientific Chemicals
04380806
5 g
Each for $778.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23210-22-2
223.275
DJKASMOURATDIE-UHFFFAOYSA-N
4990835
CC1C2=CC=CC=C2N(C1=O)C3=CC=CC=C3
C15H13NO
MFCD00798609
3-methyl-1-phenylindolin-2-one, 3-methyl-1-phenylindoline-2-one, 3-methyl-1-phenyl-2,3-dihydro-1h-indol-2-one, acmc-1ciq4, 1-phenyl-3-methylindoline-2-one, 3-methyl-1-phenyl-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one,1,3-dihydro-3-methyl-1-phenyl
3-methyl-1-phenyl-3H-indol-2-one

Specifications

23210-22-2
MFCD00798609
3-methyl-1-phenylindolin-2-one, 3-methyl-1-phenylindoline-2-one, 3-methyl-1-phenyl-2,3-dihydro-1h-indol-2-one, acmc-1ciq4, 1-phenyl-3-methylindoline-2-one, 3-methyl-1-phenyl-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one,1,3-dihydro-3-methyl-1-phenyl
CC1C2=CC=CC=C2N(C1=O)C3=CC=CC=C3
223.275
223.27
Powder
C15H13NO
1 g
DJKASMOURATDIE-UHFFFAOYSA-N
3-methyl-1-phenyl-3H-indol-2-one
4990835
97%
3-Methyl-1-phenylindolin-2-one
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only