missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenylindolin-2-one, 97%

Catalog No. AA4380803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4380803 1 g
AA4380806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4380803 Supplier Thermo Scientific Chemicals Supplier No. 04380803

CAS: 23210-22-2 | C15H13NO | 223.275 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23210-22-2
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD00798609
InChI Key DJKASMOURATDIE-UHFFFAOYSA-N
Synonym 3-methyl-1-phenylindolin-2-one, 3-methyl-1-phenylindoline-2-one, 3-methyl-1-phenyl-2,3-dihydro-1h-indol-2-one, acmc-1ciq4, 1-phenyl-3-methylindoline-2-one, 3-methyl-1-phenyl-1,3-dihydro-2h-indol-2-one, 2h-indol-2-one,1,3-dihydro-3-methyl-1-phenyl
PubChem CID 4990835
IUPAC Name 3-methyl-1-phenyl-3H-indol-2-one
SMILES CC1C2=CC=CC=C2N(C1=O)C3=CC=CC=C3

Specifications

Quantity 1 g
Formula Weight 223.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Methyl-1-phenylindolin-2-one
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only