missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-penten-4-yn-3-ol 98.0+%, TCI America™

Catalog No. p-7132657
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13125ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13125ML Supplier TCI America Supplier No. M13125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3230-69-1
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD01709006
InChI Key VBATUBQIYXCZPA-UHFFFAOYNA-N
Synonym 3-methyl-1-penten-4-yn-3-ol, 1-penten-4-yn-3-ol, 3-methyl, ethynyl methyl vinyl carbinol, 3-methyl-pent-4-en-1-yn-3-ol, 3-methyl pent-4-en-l-yn-3-0l, acmc-209hsd, 4-01-00-02309 beilstein handbook reference, ksc496k7h, wln: 1uu1xq1&1u1, 3-methyl-4-penten-1-yn-3-ol
PubChem CID 97815
IUPAC Name 3-methylpent-1-en-4-yn-3-ol
SMILES CC(O)(C=C)C#C

Specifications

Color Yellow
Boiling Point 121°C
Quantity 5 mL
UN Number 1987
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-penten-4-yn-3-ol
RTECSNumber SB4950000
TSCA No