missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-hexanol 97.0+%, TCI America™

Catalog No. M12461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12461G 1 g
M12465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12461G Supplier TCI America Supplier No. M12461G
Only null left

Chemical Identifiers

CAS 13231-81-7
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD01321178
InChI Key YGZVAQICDGBHMD-UHFFFAOYSA-N
PubChem CID 25793
IUPAC Name 3-methylhexan-1-ol
SMILES CCCC(C)CCO

Specifications

Color Colorless
Boiling Point 96°C
Quantity 1 g
Formula Weight 116.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-hexanol
EINECSNumber (2)-0217
TSCA No