missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butene (ca. 12.5% in Tetrahydrofuran, ca. 1.5mol/L), TCI America™

Supplier:  TCI America M2565100ML

Catalog No. M2565100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methyl-1-butene (ca. 12.5% in Tetrahydrofuran, ca. 1.5mol/L)
Yellow
MFCD00008937
1993
YHQXBTXEYZIYOV-UHFFFAOYSA-N
3-methylbut-1-ene
11239
70.14
563-45-1
C5H10
100 mL
Isoamylene, Isopentene, Isopropylethylene
CC(C)C=C
70.14
CHEBI:77914
Liquid

Chemical Identifiers

563-45-1
70.14
YHQXBTXEYZIYOV-UHFFFAOYSA-N
11239
3-methylbut-1-ene
C5H10
MFCD00008937
Isoamylene, Isopentene, Isopropylethylene
CHEBI:77914
CC(C)C=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0019

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions