missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol 99.0+%, TCI America™

Catalog No. p-7131310
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I028925ML 25 mL
I0289500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I028925ML Supplier TCI America Supplier No. I028925ML
Only null left

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 1105
Formula Weight 88.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-butanol
EINECSNumber (2)-0217
RTECSNumber EL5425000
TSCA Yes