missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, mixture of isomers, 99%

CAS: 123-51-3 | C5H12O | 88.15 g/mol

Supplier:  Thermo Scientific Chemicals L13660AP

 View more versions of this product

Catalog No. AAL13660AP


Only null left
Explore available promotions

Description

Description

3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac′s reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

Solubility
Miscible with water, alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, acetone and oils.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides and acid anhydrides.
Specifications

Specifications

3-Methyl-1-butanol
123-51-3
0.809
45°C (113°F)
C5H12O
MFCD00002934
UN1105
14,5195
Miscible with water,alcohol,ether,benzene,chloroform,petroleum ether,glacial acetic acid,acetone and oils.
CC(C)CCO
88.15
CHEBI:15837
99%
mixture of isomers
-117°C
130°C to 132°C
Unpleasant
1.406
500 mL
1718835
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methylbutan-1-ol
31260
88.15

Chemical Identifiers

123-51-3
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
31260
3-methylbutan-1-ol
C5H12O
MFCD00002934
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
CHEBI:15837
CC(C)CCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS

EINECSNumber : 204-633-5

RTECSNumber : EL5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only