missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, ACS reagent

CAS: 123-51-3 | C5H12O | 88.15 g/mol

Supplier:  Thermo Scientific Chemicals 412730010

 View more versions of this product

Catalog No. AC412730010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methyl-1-butanol
123-51-3
0.8000g/mL
45°C
C5H12O
(CH3)2CHCH2CH2OH
1 L
0.003% max.
15, 5241
Solubility in water: 25g/L (20°C). Other solubilities: miscible with alcohol,ether,benzene,oils,,chloroform
CC(C)CCO
88.15
4.3 mPa.s (20°C)
88.15
ACS Reagent
Liquid
98.5%
-70.0°C
130.0°C
Glass bottle
1.4050 to 1.4070
MFCD00002934
01, 392
0.8
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methylbutan-1-ol
31260
CHEBI:15837
≥98.5%
0.002 meq/g max

Chemical Identifiers

123-51-3
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
31260
3-methylbutan-1-ol
C5H12O
MFCD00002934
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
CHEBI:15837
CC(C)CCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin

GHS Signal Word: Warning

EINECSNumber : 204-633-5

RTECSNumber : EL5425000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only