missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,2-cyclopentanedione 98.0+%, TCI America™

Catalog No. M290425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M290425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M290425G Supplier TCI America Supplier No. M290425G

Chemical Identifiers

CAS 765-70-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001417
InChI Key OACYKCIZDVVNJL-UHFFFAOYSA-N
Synonym 3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
PubChem CID 61209
IUPAC Name 3-methylcyclopentane-1,2-dione
SMILES CC1CCC(=O)C1=O

Specifications

Melting Point 106°C
Quantity 25 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-1,2-cyclopentanedione
EINECSNumber (3)-2406
TSCA Yes