missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,2-butadiene 97.0+%, TCI America™

Catalog No. m06211ml
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06211ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M06211ML Supplier TCI America Supplier No. M06211ML

Chemical Identifiers

CAS 598-25-4
Molecular Formula C5H8
Molecular Weight (g/mol) 68.12
MDL Number MFCD00008896
InChI Key PAKGDPSCXSUALC-UHFFFAOYSA-N
Synonym 3-methyl-1,2-butadiene, 1,1-dimethylallene, 1,2-butadiene, 3-methyl, 3,3-dimethylallene, 1,1-dimethylallylene, 2-methyl-2,3-butadiene, acmc-1aqbc, ch2=c=c ch3 2, 3-methyl-1,2-butadiene 1g
PubChem CID 11714
IUPAC Name 3-methylbuta-1,2-diene
SMILES CC(C)=C=C

Specifications

Color Colorless
Boiling Point 40°C
Quantity 1 mL
UN Number 3295
Formula Weight 68.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-1,2-butadiene
EINECSNumber (2)-0020
TSCA No
Recommended Storage Refrigerator