missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythiophene 98.0+%, TCI America™

Catalog No. m109325g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
M109325G 25 g
M10931G 1 g
M10935G 5 g
3 options

Catalog No. M109325G

Supplier: TCI America M109325G

Only null left

Chemical Identifiers

CAS 17573-92-1
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.162
MDL Number MFCD00043894
InChI Key RFSKGCVUDQRZSD-UHFFFAOYSA-N
Synonym thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411
PubChem CID 140249
IUPAC Name 3-methoxythiophene
SMILES COC1=CSC=C1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 25 g
UN Number 1993
Formula Weight 114.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxythiophene
EINECSNumber (5)-6392
TSCA No
Recommended Storage Refrigerator