missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxythiophene 98.0+%, TCI America™
Supplier: TCI America M10931G
Specifications
| 3-Methoxythiophene | |
| Yellow | |
| C5H6OS | |
| 1 g | |
| thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411 | |
| COC1=CSC=C1 | |
| 114.162 | |
| 114.16 | |
| Liquid |
| 17573-92-1 | |
| 82°C | |
| MFCD00043894 | |
| 1993 | |
| RFSKGCVUDQRZSD-UHFFFAOYSA-N | |
| 3-methoxythiophene | |
| 140249 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 17573-92-1 | |
| 114.162 | |
| RFSKGCVUDQRZSD-UHFFFAOYSA-N | |
| 140249 | |
| COC1=CSC=C1 |
| C5H6OS | |
| MFCD00043894 | |
| thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411 | |
| 3-methoxythiophene |
Safety and Handling
EINECSNumber : (5)-6392
TSCA : No
Recommended Storage : Refrigerator