missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythiophene, 99%

Catalog No. AAB2194006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2194006 5 g
AAB2194014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2194006 Supplier Thermo Scientific Chemicals Supplier No. B2194006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17573-92-1 | C5H6OS | 114.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17573-92-1
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.162
MDL Number MFCD00043894
InChI Key RFSKGCVUDQRZSD-UHFFFAOYSA-N
Synonym thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411
PubChem CID 140249
IUPAC Name 3-methoxythiophene
SMILES COC1=CSC=C1

Specifications

Density 1.147
Boiling Point 60°C to 61°C (12 mmHg)
Flash Point 49°C (120°F)
Odor Recognizable
Refractive Index 1.532
Quantity 5 g
UN Number UN3271
Beilstein 106404
Sensitivity Air sensitive
Formula Weight 114.17
Percent Purity 99%
Chemical Name or Material 3-Methoxythiophene
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only