missing translation for 'onlineSavingsMsg'
Learn More

(3-methoxythien-2-yl)methylamine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 946409-37-6
Molecular Formula C6H9NOS
Molecular Weight (g/mol) 143.20
MDL Number MFCD11841071
InChI Key GDDVAKBOBBYCLK-UHFFFAOYSA-N
Synonym 3-methoxythien-2-yl methylamine, 3-methoxythiophen-2-yl methanamine, 2-aminomethyl-3-methoxythiophene, 1-3-methoxythiophen-2-yl methanamine, 3-methoxy-2-thienyl methylamine, 3-methoxy-2-thiophenyl methanamine, 3-methoxy-2-thienyl methyl amine
PubChem CID 43811049
IUPAC Name (3-methoxythiophen-2-yl)methanamine
SMILES COC1=C(CN)SC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC68613CB
SDS View Documents
Thermo Scientific Chemicals
CC68613CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC68613DA
SDS View Documents
Thermo Scientific Chemicals
CC68613DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

946409-37-6
143.20
GDDVAKBOBBYCLK-UHFFFAOYSA-N
43811049
COC1=C(CN)SC=C1
C6H9NOS
MFCD11841071
3-methoxythien-2-yl methylamine, 3-methoxythiophen-2-yl methanamine, 2-aminomethyl-3-methoxythiophene, 1-3-methoxythiophen-2-yl methanamine, 3-methoxy-2-thienyl methylamine, 3-methoxy-2-thiophenyl methanamine, 3-methoxy-2-thienyl methyl amine
(3-methoxythiophen-2-yl)methanamine

Specifications

946409-37-6
100.0
Amber Glass Bottle
MFCD11841071
3-methoxythien-2-yl methylamine, 3-methoxythiophen-2-yl methanamine, 2-aminomethyl-3-methoxythiophene, 1-3-methoxythiophen-2-yl methanamine, 3-methoxy-2-thienyl methylamine, 3-methoxy-2-thiophenyl methanamine, 3-methoxy-2-thienyl methyl amine
COC1=C(CN)SC=C1
143.20
143.21
(3-Methoxythien-2-yl)methylamine
94.5
95%
C6H9NOS
250mg
GDDVAKBOBBYCLK-UHFFFAOYSA-N
(3-methoxythiophen-2-yl)methanamine
43811049
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'