missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyquinoline-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. p-7035976
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
AAH5421103 1 g
1 options

Catalog No. AAH5421103

Supplier: Thermo Scientific Chemicals H5421103

Only null left

3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 881656-35-5
Molecular Formula C16H20BNO3
Molecular Weight (g/mol) 285.15
MDL Number MFCD20527212
InChI Key MGEKTRDPBDREKY-UHFFFAOYSA-N
Synonym 3-methoxyquinoline-5-boronic acid pinacol ester, 3-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-quinoline, 3-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 3-methoxy-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl-quinoline
PubChem CID 68783972
IUPAC Name 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
SMILES COC1=CN=C2C=CC=C(B3OC(C)(C)C(C)(C)O3)C2=C1

Specifications

Quantity 1 g
Formula Weight 285.15
Percent Purity 96%
Chemical Name or Material 3-Methoxyquinoline-5-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only