missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropylamine 99.0+%, TCI America™

Catalog No. p-7132008
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M012725ML 25 mL
M0127500ML 500 mL
2 options

Catalog No. M012725ML

Supplier: TCI America M012725ML

Only null left

Chemical Identifiers

CAS 5332-73-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00014831
InChI Key FAXDZWQIWUSWJH-UHFFFAOYSA-N
Synonym 3-methoxypropylamine, 1-propanamine, 3-methoxy, 1-amino-3-methoxypropane, 3-aminopropyl methyl ether, 3-methyoxypropylamine, 3-methoxy-n-propylamine, 3-methoxy-1-propanamine, propylamine, 3-methoxy, 3-mpa, propanolamine methyl ether
PubChem CID 1672
IUPAC Name 3-methoxypropan-1-amine
SMILES COCCCN

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 2734
Formula Weight 89.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxypropylamine
EINECSNumber (2)-0385
RTECSNumber UI3335000
TSCA Yes