missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 257520010

 View more versions of this product

Catalog No. AC257520010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methoxypropylamine
5332-73-0
100.0
Colorless
118°C
Authentic
Glass bottle
1.4165 to 1.4185
MFCD00014831
0.87
Solubility in water: miscible. Other solubilities: miscible with ethanol,toluene,acetone,ether,,carbon tetrachloride and hexane
COCCCN
89.14
0.91 mPa.s (20°C)
99+%
99+%
99.0
-65°C
0.8700g/mL
48°C
99% min. (GC)
C4H11NO
CH3OCH2CH2CH2NH2
1 L
3-methoxypropylamine, 1-propanamine, 3-methoxy, 1-amino-3-methoxypropane, 3-aminopropyl methyl ether, 3-methyoxypropylamine, 3-methoxy-n-propylamine, 3-methoxy-1-propanamine, propylamine, 3-methoxy, 3-mpa, propanolamine methyl ether
FAXDZWQIWUSWJH-UHFFFAOYSA-N
3-methoxypropan-1-amine
1672
89.12
Liquid

Chemical Identifiers

5332-73-0
89.14
FAXDZWQIWUSWJH-UHFFFAOYSA-N
1672
COCCCN
C4H11NO
MFCD00014831
3-methoxypropylamine, 1-propanamine, 3-methoxy, 1-amino-3-methoxypropane, 3-aminopropyl methyl ether, 3-methyoxypropylamine, 3-methoxy-n-propylamine, 3-methoxy-1-propanamine, propylamine, 3-methoxy, 3-mpa, propanolamine methyl ether
3-methoxypropan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 226-241-3

RTECSNumber : UI3335000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only