missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropionitrile 99.0+%, TCI America™

Catalog No. p-7132283
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M065325ML 25 mL
M0653500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M065325ML Supplier TCI America Supplier No. M065325ML
Only null left

Chemical Identifiers

CAS 110-67-8
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00001958
InChI Key OOWFYDWAMOKVSF-UHFFFAOYSA-N
Synonym 3-methoxypropionitrile, propanenitrile, 3-methoxy, 2-cyanoethyl methyl ether, 3-methoxypropylnitrile, beta-methoxypropionitrile, 1-cyano-2-methoxyethane, propionitrile, 3-methoxy, 3-methoxypropannitril, 3-methoxypropiononitrile, beta-methyoxypropionitrile
PubChem CID 61032
IUPAC Name 3-methoxypropanenitrile
SMILES COCCC#N

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 166°C
Quantity 25 mL
Formula Weight 85.11
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxypropionitrile
EINECSNumber (2)-1536
RTECSNumber TZ4870000
TSCA Yes