missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropionitrile, 99%

Catalog No. AAL07055AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL07055AP 500 mL
AAL07055AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL07055AP Supplier Thermo Scientific Chemicals Supplier No. L07055AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-67-8 | C4H7NO | 85.106 g/mol

3-Methoxypropionitrile is used as a component in high power lithium ion batteries. It is used as pharmaceutical intermediates. It acts as a good solvent for plastic polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxypropionitrile is used as a component in high power lithium ion batteries. It is used as pharmaceutical intermediates. It acts as a good solvent for plastic polymers.

Solubility
Soluble in water. (540 g/L) at 25°C.

Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 110-67-8
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00001958
InChI Key OOWFYDWAMOKVSF-UHFFFAOYSA-N
Synonym 3-methoxypropionitrile, propanenitrile, 3-methoxy, 2-cyanoethyl methyl ether, 3-methoxypropylnitrile, beta-methoxypropionitrile, 1-cyano-2-methoxyethane, propionitrile, 3-methoxy, 3-methoxypropannitril, 3-methoxypropiononitrile, beta-methyoxypropionitrile
PubChem CID 61032
IUPAC Name 3-methoxypropanenitrile
SMILES COCCC#N

Specifications

Melting Point -63°C
Density 0.938
pH 6.8
Boiling Point 164°C to 165°C
Flash Point 66°C (150°F)
Odor Weak
Refractive Index 1.403
Quantity 500 mL
Beilstein 1739284
Solubility Information Soluble in water. (540g/L) at 25°C.
Formula Weight 85.11
Percent Purity 99%
Chemical Name or Material 3-Methoxypropionitrile
Show More Show Less
Hazard Category H227-H319
Hazard Statement GHS H Statement
H319-H335-H227
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 203-790-7
RTECSNumber TZ4870000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only