missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropionitrile, 98%

Catalog No. AC257440025
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC257440025 2.5 L Glass bottle
AC257441000 100 mL Glass bottle
AC257445000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC257440025 Supplier Thermo Scientific Chemicals Supplier No. 257440025
Only null left

CAS: 110-67-8 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-67-8
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00001958
InChI Key OOWFYDWAMOKVSF-UHFFFAOYSA-N
Synonym 3-methoxypropionitrile, propanenitrile, 3-methoxy, 2-cyanoethyl methyl ether, 3-methoxypropylnitrile, beta-methoxypropionitrile, 1-cyano-2-methoxyethane, propionitrile, 3-methoxy, 3-methoxypropannitril, 3-methoxypropiononitrile, beta-methyoxypropionitrile
PubChem CID 61032
IUPAC Name 3-methoxypropanenitrile
SMILES COCCC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -62.9°C
Color Colorless to Yellow
Density 0.9380g/mL
Boiling Point 164.0°C to 165.0°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4020 to 1.4040
Linear Formula CH3OCH2CH2CN
Quantity 2.5 L
Specific Gravity 0.938
Solubility Information Solubility in water: 335g/L (20°C)
Formula Weight 85.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxypropionitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 203-790-7
RTECSNumber TZ4870000
TSCA TSCA

RUO – Research Use Only