missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphthalide 98.0+%, TCI America™

Catalog No. M20075G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20075G Supplier TCI America Supplier No. M20075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4122-57-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
InChI Key OIIJJGAFRGJQSQ-UHFFFAOYSA-N
Synonym 3-Methoxy-1(3H)-isobenzofuranone
PubChem CID 349017
IUPAC Name 3-methoxy-3H-2-benzofuran-1-one
SMILES COC1C2=CC=CC=C2C(=O)O1

Specifications

Melting Point 45°C
Color White
Boiling Point 146°C
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxyphthalide
TSCA No
Recommended Storage Refrigerator