missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M13221G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13221G Supplier TCI America Supplier No. M13221G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10365-98-7
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00161359
InChI Key NLLGFYPSWCMUIV-UHFFFAOYSA-N
Synonym 3-methoxyphenyl boronic acid, 3-methoxybenzeneboronic acid, 3-methoxyphenyl boranediol, 3-methoxypenylboronic acid, 3-boronanisole, 3-boronoanisole, boronic acid, 3-methoxyphenyl, m-methoxyphenylboronic acid, 3-methoxy phenylboronic acid
PubChem CID 2734370
IUPAC Name (3-methoxyphenyl)boronic acid
SMILES COC1=CC=CC(=C1)B(O)O

Specifications

Melting Point 160°C
Quantity 1 g
Formula Weight 151.96
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxyphenylboronic Acid (contains varying amounts of Anhydride)